2-Chlor-4-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Chloro-4-methylbenzeneboronic acid

BB-2194-5G BB-2194-1G BB-2194-25G
COBBBB-2194-5GEA 63.4 EUR
COBBBB-2194-5G COBBBB-2194-1G COBBBB-2194-25G
2-Chlor-4-methylphenylboronsäure
2-Chlor-4-methylphenylboronsäure
Formel: C₇H₈BClO₂
Molecular Weight: 170.40 g/mol
MDL: MFCD03411936
CAS-Nummer: 145349-62-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR