4-Fluor-2-methylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-4-fluorophenylboronic acid
BB-2204-5G
BB-2204-1G
BB-2204-25G
COBBBB-2204-5GEA
55.9
EUR
COBBBB-2204-5G
COBBBB-2204-1G
COBBBB-2204-25G
4-Fluor-2-methylphenylboronsäure
4-Fluor-2-methylphenylboronsäure
Formel:
C₇H₈BFO₂ Molecular Weight: 153.95 g/mol |
MDL:
MFCD02093072 CAS-Nummer: 139911-29-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...