4-Fluor-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-4-fluorophenylboronic acid

BB-2204-5G BB-2204-1G BB-2204-25G
COBBBB-2204-5GEA 55.9 EUR
COBBBB-2204-5G COBBBB-2204-1G COBBBB-2204-25G
4-Fluor-2-methylphenylboronsäure
4-Fluor-2-methylphenylboronsäure
Formel: C₇H₈BFO₂
Molecular Weight: 153.95 g/mol
MDL: MFCD02093072
CAS-Nummer: 139911-29-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR