4-Methoxy-2-methylphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-Methoxy-2-methylbenzeneboronic acid, (4-Methoxy-2-methylphenyl)boronic acid
BB-2206-25G
BB-2206-1G
BB-2206-5G
COBBBB-2206-25GEA
310
EUR
COBBBB-2206-25G
COBBBB-2206-1G
COBBBB-2206-5G
4-Methoxy-2-methylphenylboronsäure
4-Methoxy-2-methylphenylboronsäure
Formel:
C₈H₁₁BO₃ Molecular Weight: 165.99 g/mol |
MDL:
MFCD02684315 CAS-Nummer: 208399-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...