4-Methoxy-2-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Methoxy-2-methylbenzeneboronic acid, (4-Methoxy-2-methylphenyl)boronic acid

BB-2206-25G BB-2206-1G BB-2206-5G
COBBBB-2206-25GEA 310 EUR
COBBBB-2206-25G COBBBB-2206-1G COBBBB-2206-5G
4-Methoxy-2-methylphenylboronsäure
4-Methoxy-2-methylphenylboronsäure
Formel: C₈H₁₁BO₃
Molecular Weight: 165.99 g/mol
MDL: MFCD02684315
CAS-Nummer: 208399-66-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR