1-Boc-5-cyan-2-indolboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(1-(tert-Butoxycarbonyl)-5-cyano-1H-indol-2-yl)boronic acid, 5-Cyano-1H-indole-2-boronic acid, N-BOC protected
BB-2211-1G
COBBBB-2211-1GEA
335
EUR
COBBBB-2211-1G
1-Boc-5-cyan-2-indolboronsäure
1-Boc-5-cyan-2-indolboronsäure
Formel:
C₁₄H₁₅BN₂O₄ Molecular Weight: 286,09 g/mol |
MDL:
MFCD04114578 CAS-Nummer: 475102-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...