3-Acetamido-4-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2218-250M
COBBBB-2218-250MEA 0 EUR
COBBBB-2218-250M
3-Acetamido-4-nitrophenylboronsäure
3-Acetamido-4-nitrophenylboronsäure
Formel: C₈H₉BN₂O₅
MDL: MFCD12025959
CAS-Nummer: 78887-37-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR