5-Acetamido-2-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-Acetamido-2-nitro)benzeneboronicacid

BB-2219-1G
COBBBB-2219-1GEA 451 EUR
COBBBB-2219-1G
5-Acetamido-2-nitrophenylboronsäure
5-Acetamido-2-nitrophenylboronsäure
Formel: C₈H₉BN₂O₅
Molecular Weight: 223,9800 g/mol
MDL: MFCD06659829
CAS-Nummer: 78887-36-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR