3-Aminophenylboronsäure Hydrochlorid
Lieferant: COMBI-BLOCKS
BB-2229-5G
BB-2229-25G
BB-2229-1G
COBBBB-2229-5GEA
0
EUR
COBBBB-2229-5G
COBBBB-2229-25G
COBBBB-2229-1G
3-Aminophenylboronsäure Hydrochlorid
3-Aminophenylboronsäure Hydrochlorid
Formel:
C₆H₉BClNO₂ |
MDL:
MFCD00191748 CAS-Nummer: 85006-23-1 EINECS: 285-052-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...