2-Benzyloxy-5-methylphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-(Benzyloxy)-5-methylphenyl)boronic acid

BB-2338-1G
COBBBB-2338-1GEA 114 EUR
COBBBB-2338-1G
2-Benzyloxy-5-methylphenylboronsäure
2-Benzyloxy-5-methylphenylboronsäure
Formel: C₁₄H₁₅BO₃
Molecular Weight: 242,0800 g/mol
MDL: MFCD06801733
CAS-Nummer: 127972-17-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR