5-Brom-2-chlor-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Brom-2-chlorpyridin-4-boronsäure, 5-Brom-2-chlor-4-pyridinylboronsäure

BB-2361-1G
COBBBB-2361-1GEA 451 EUR
COBBBB-2361-1G
5-Brom-2-chlor-4-pyridinboronsäure
5-Bromo-2-chloro-4-pyridineboronic acid
Formel: C₅H₄BBrClNO₂
Molecular Weight: 236,26 g/mol
MDL: MFCD07783852
CAS-Nummer: 871329-63-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR