N-tert-Butyl-3-boronobenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-[N-(tert-Butyl)sulphamoyl]phenylboronic Acid

BB-2408-1G
COBBBB-2408-1GEA 358 EUR
COBBBB-2408-1G
N-tert-Butyl-3-boronobenzolsulfonamid
N-tert-Butyl-3-boronobenzolsulfonamid
Formel: C₁₀H₁₆BNO₄S
Molecular Weight: 257,1143 g/mol
MDL: MFCD06659842
CAS-Nummer: 221290-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR