N-Cyclohexyl-4-boronobenzolsulfonamid
Lieferant: COMBI-BLOCKS
BB-2429-1G
BB-2429-5G
COBBBB-2429-1GEA
0
EUR
COBBBB-2429-1G
COBBBB-2429-5G
N-Cyclohexyl-4-boronobenzolsulfonamid
N-Cyclohexyl-4-boronobenzolsulfonamid
Formel:
C₁₂H₁₈BNO₄S |
MDL:
MFCD07363754 CAS-Nummer: 871329-66-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...