4-Isobutylphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2441-100G
BB-2441-5G
BB-2441-1G
BB-2441-25G
COBBBB-2441-100GEA
0
EUR
COBBBB-2441-100G
COBBBB-2441-5G
COBBBB-2441-1G
COBBBB-2441-25G
4-Isobutylphenylboronsäure
4-Isobutylphenylboronsäure
Formel:
C₁₀H₁₅BO₂ |
MDL:
MFCD01632198 CAS-Nummer: 153624-38-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...