2-Fluor-3-methoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Fluoro-3-methoxybenzeneboronic acid, (2-Fluoro-3-methoxyphenyl)boronic acid
BB-2460-5G
BB-2460-1G
COBBBB-2460-5GEA
44.8
EUR
COBBBB-2460-5G
COBBBB-2460-1G
2-Fluor-3-methoxyphenylboronsäure
2-Fluor-3-methoxyphenylboronsäure
Formel:
C₇H₈BFO₃ Molecular Weight: 169,95 g/mol |
MDL:
MFCD02094728 CAS-Nummer: 352303-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...