4-Formyl-1-naphthalinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Formylnaphthalen-1-yl)boronic acid

BB-2465-1G
COBBBB-2465-1GEA 1410 EUR
COBBBB-2465-1G
4-Formyl-1-naphthalinboronsäure
4-Formyl-1-naphthalinboronsäure
Formel: C₁₁H₉BO₃
Molecular Weight: 200.00 g/mol
MDL: MFCD01632205
CAS-Nummer: 332398-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR