3-(Brommethyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Dihydroxy(3-(brommethyl)phenyl)boran, (3-(Brommethyl)phenyl)metaborsäure, 3-(Brommethyl)benzolboronsäure

BB-2468-5G BB-2468-25G BB-2468-1G
COBBBB-2468-5GEA 0 EUR
COBBBB-2468-5G COBBBB-2468-25G COBBBB-2468-1G
3-(Brommethyl)phenylboronsäure
3-(Brommethyl)phenylboronsäure
Formel: C₇H₈BBrO₂
MDL: MFCD01632207
CAS-Nummer: 51323-43-4
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR