1-Cyclopentenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: Cyclopent-1-en-1-ylboronic acid

BB-2510-1G
COBBBB-2510-1GEA 208 EUR
COBBBB-2510-1G
1-Cyclopentenylboronsäure
1-Cyclopentenylboronsäure
Formel: C₅H₉BO₂
Molecular Weight: 111,94 g/mol
MDL: MFCD02179496
CAS-Nummer: 850036-28-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR