4,4-Dimethylcyclohexa-1,5-dienylboronsäure Mononatriumsalz

Lieferant: COMBI-BLOCKS

BB-2534-1G
COBBBB-2534-1GEA 0 EUR
COBBBB-2534-1G
4,4-Dimethylcyclohexa-1,5-dienylboronsäure Mononatriumsalz
4,4-Dimethylcyclohexa-1,5-dienylboronsäure Mononatriumsalz
Formel: C₈H₁₂BNaO₂
MDL: MFCD07363759
CAS-Nummer: 871329-70-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR