4-Methylcyclohexen-1-ylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Methylcyclohex-1-en-1-yl)boronic acid, 4-Methyl-1-cyclohexen-1-ylboronic acid

BB-2538-1G
COBBBB-2538-1GEA 692 EUR
COBBBB-2538-1G
4-Methylcyclohexen-1-ylboronsäure
4-Methylcyclohexene-1-ylboronic acid
Formel: C₇H₁₃BO₂
Molecular Weight: 139,99 g/mol
MDL: MFCD06659857
CAS-Nummer: 850567-92-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR