3-Formyl-2-thiophenboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Formylthiophene-2-boronic acid, (3-Formylthiophen-2-yl)boronic acid

BB-2550-1G
COBBBB-2550-1GEA 114 EUR
COBBBB-2550-1G
3-Formyl-2-thiophenboronsäure
3-Formyl-2-thiophenboronsäure
Formel: C₅H₅BO₃S
Molecular Weight: 155,97 g/mol
MDL: MFCD01075679
CAS-Nummer: 17303-83-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR