2-Formyl-3-thiophenboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Formylthiophene-3-boronic acid, (2-Formylthiophen-3-yl)boronic acid

BB-2552-1G
COBBBB-2552-1GEA 159 EUR
COBBBB-2552-1G
2-Formyl-3-thiophenboronsäure
2-Formyl-3-thiophenboronsäure
Formel: C₅H₅BO₃S
Molecular Weight: 155,97 g/mol
MDL: MFCD01075678
CAS-Nummer: 4347-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR