5-Oxocyclopenten-1-ylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Oxocyclopentene-1-boronic acid

BB-2564-250M
COBBBB-2564-250MEA 909 EUR
COBBBB-2564-250M
5-Oxocyclopenten-1-ylboronsäure
5-Oxocyclopenten-1-ylboronic acid
Formel: C₅H₇BO₃
Molecular Weight: 125,92 g/mol
MDL: MFCD08056354
CAS-Nummer: 871329-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR