4-(Methylsulfonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Methylsulphonyl)benzeneboronic acid, 4-(Methanesulphonyl)phenylboronic acid

BB-2580-25G BB-2580-1G BB-2580-100G BB-2580-5G
COBBBB-2580-25GEA 0 EUR
COBBBB-2580-25G COBBBB-2580-1G COBBBB-2580-100G COBBBB-2580-5G
4-(Methylsulfonyl)phenylboronsäure
4-(Methylsulfonyl)phenylboronsäure
Formel: C₇H₉BO₄S
Molecular Weight: 200.02 g/mol
MDL: MFCD01630820
CAS-Nummer: 149104-88-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR