2-Chlor-3-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Chloro-3-methoxyphenyl)boronic acid

BB-2581-1G BB-2581-250M
COBBBB-2581-1GEA 221 EUR
COBBBB-2581-1G COBBBB-2581-250M
2-Chlor-3-methoxyphenylboronsäure
2-Chlor-3-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
Molecular Weight: 186,4006 g/mol
MDL: MFCD12913933
CAS-Nummer: 854778-30-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR