3-Chlor-5-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-5-methoxybenzeneboronic acid

BB-2589-1G
COBBBB-2589-1GEA 96.3 EUR
COBBBB-2589-1G
3-Chlor-5-methoxyphenylboronsäure
3-Chlor-5-methoxyphenylboronsäure
Formel: C₇H₈BClO₃
Molecular Weight: 186,40 g/mol
MDL: MFCD03095046
CAS-Nummer: 915201-07-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR