2,6-Dibrom-3-methoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2594-1G BB-2594-5G
COBBBB-2594-1GEA 0 EUR
COBBBB-2594-1G COBBBB-2594-5G
2,6-Dibrom-3-methoxyphenylboronsäure
2,6-Dibrom-3-methoxyphenylboronsäure
Formel: C₇H₇BBr₂O₃
MDL: MFCD06659861
CAS-Nummer: 850567-93-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR