3-(Methylsulfonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2626-5G BB-2626-25G BB-2626-1G
COBBBB-2626-5GEA 0 EUR
COBBBB-2626-5G COBBBB-2626-25G COBBBB-2626-1G
3-(Methylsulfonyl)phenylboronsäure
3-(Methylsulfonyl)phenylboronsäure
Formel: C₇H₉BO₄S
MDL: MFCD03092935
CAS-Nummer: 373384-18-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR