3-(Methylsulfonyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2626-5G
BB-2626-25G
BB-2626-1G
COBBBB-2626-5GEA
0
EUR
COBBBB-2626-5G
COBBBB-2626-25G
COBBBB-2626-1G
3-(Methylsulfonyl)phenylboronsäure
3-(Methylsulfonyl)phenylboronsäure
Formel:
C₇H₉BO₄S |
MDL:
MFCD03092935 CAS-Nummer: 373384-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...