2-(Trifluormethoxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2650-1G BB-2650-5G BB-2650-25G BB-2650-100G
COBBBB-2650-1GEA 0 EUR
COBBBB-2650-1G COBBBB-2650-5G COBBBB-2650-25G COBBBB-2650-100G
2-(Trifluormethoxy)phenylboronsäure
2-(Trifluormethoxy)phenylboronsäure
Formel: C₇H₆BF₃O₃
MDL: MFCD01320763
CAS-Nummer: 175676-65-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR