2-(Trifluormethoxy)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-2650-1G
BB-2650-5G
BB-2650-25G
BB-2650-100G
COBBBB-2650-1GEA
0
EUR
COBBBB-2650-1G
COBBBB-2650-5G
COBBBB-2650-25G
COBBBB-2650-100G
2-(Trifluormethoxy)phenylboronsäure
2-(Trifluormethoxy)phenylboronsäure
Formel:
C₇H₆BF₃O₃ |
MDL:
MFCD01320763 CAS-Nummer: 175676-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...