2,6-Difluorphenylboronsäure

Lieferant: COMBI-BLOCKS
Achtung

Synonyme: Dihydroxy(2,6-difluorophenyl)borane, 2,6-Difluorobenzeneboronic acid, (2,6-Difluorophenyl)metaboric acid

BB-2659-100G BB-2659-5G BB-2659-1G BB-2659-25G
COBBBB-2659-100GEA 385 EUR
COBBBB-2659-100G COBBBB-2659-5G COBBBB-2659-1G COBBBB-2659-25G
2,6-Difluorphenylboronsäure
2,6-Difluorphenylboronsäure
Formel: C₆H₅BF₂O₂
Molecular Weight: 157.91 g/mol
MDL: MFCD00792436
CAS-Nummer: 162101-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR