2,5-Dichlorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2662-25G BB-2662-1G BB-2662-5G
COBBBB-2662-25GEA 242 EUR
COBBBB-2662-25G COBBBB-2662-1G COBBBB-2662-5G
2,5-Dichlorphenylboronsäure
2,5-Dichlorphenylboronsäure
Formel: C₆H₅BCl₂O₂
Molecular Weight: 190.82 g/mol
MDL: MFCD01863182
CAS-Nummer: 135145-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR