4-(Trifluormethyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-(Trifluormethyl)phenyl)metaborsäure, Dihydroxy(4-(trifluormethyl)phenyl)boran, 4-(Trifluormethyl)benzolboronsäure
BB-2673-5G
BB-2673-25G
BB-2673-100G
COBBBB-2673-5GEA
25.5
EUR
COBBBB-2673-5G
COBBBB-2673-25G
COBBBB-2673-100G
4-(Trifluormethyl)phenylboronsäure
4-(Trifluormethyl)phenylboronsäure
Formel:
C₇H₆BF₃O₂ Molecular Weight: 189.93 g/mol |
MDL:
MFCD00151855 CAS-Nummer: 128796-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...