2,4,6-Trifluorphenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2674-5G BB-2674-1G
COBBBB-2674-5GEA 0 EUR
COBBBB-2674-5G COBBBB-2674-1G
2,4,6-Trifluorphenylboronsäure
2,4,6-Trifluorphenylboronsäure
Formel: C₆H₄BF₃O₂
MDL: MFCD01863169
CAS-Nummer: 182482-25-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR