2,4,6-Trimethoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
|
Achtung
|
Synonyme:
2,4,6-Trimethoxybenzeneboronic acid, (2,4,6-Trimethoxyphenyl)metaboric acid, Dihydroxy(2,4,6-trimethoxyphenyl)borane
BB-2689-1G
COBBBB-2689-1GEA
169
EUR
COBBBB-2689-1G
2,4,6-Trimethoxyphenylboronsäure
2,4,6-Trimethoxyphenylboronsäure
Formel:
C₉H₁₃BO₅ Molecular Weight: 212,01 g/mol |
MDL:
MFCD01114642 CAS-Nummer: 135159-25-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...