2-Brom-4-pyridinboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Brompyridin-4-boronsäure, 2-Brom-4-pyridinylboronsäure

BB-2708-1G
COBBBB-2708-1GEA 218 EUR
COBBBB-2708-1G
2-Brom-4-pyridinboronsäure
2-Bromo-4-pyridineboronic acid
Formel: C₅H₅BBrNO₂
Molecular Weight: 201,81 g/mol
MDL: MFCD06798244
CAS-Nummer: 458532-94-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR