5-Chlor-2-ethoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-Chloro-2-ethoxybenzeneboronic acid

BB-2712-100G BB-2712-1G BB-2712-5G BB-2712-25G
COBBBB-2712-100GEA 692 EUR
COBBBB-2712-100G COBBBB-2712-1G COBBBB-2712-5G COBBBB-2712-25G
5-Chlor-2-ethoxyphenylboronsäure
5-Chlor-2-ethoxyphenylboronsäure
Formel: C₈H₁₀BClO₃
Molecular Weight: 200.43 g/mol
MDL: MFCD03427055
CAS-Nummer: 352534-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR