5-Fluor-(2-methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2727-5G BB-2727-1G
COBBBB-2727-5GEA 0 EUR
COBBBB-2727-5G COBBBB-2727-1G
5-Fluor-(2-methoxycarbonyl)phenylboronsäure
5-Fluor-(2-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BFO₄
MDL: MFCD03095367
CAS-Nummer: 850568-05-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR