4-Fluor-(2-methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2728-1G
COBBBB-2728-1GEA 0 EUR
COBBBB-2728-1G
4-Fluor-(2-methoxycarbonyl)phenylboronsäure
4-Fluor-(2-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BFO₄
MDL: MFCD07363762
CAS-Nummer: 871329-81-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR