4-(Heptyloxy)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Heptyloxyphenylboronic acid, 4-n-Heptyloxybenzeneboronic acid, 4-Heptyloxybenzeneboronic acid

BB-2736-5G BB-2736-25G BB-2736-1G
COBBBB-2736-5GEA 0 EUR
COBBBB-2736-5G COBBBB-2736-25G COBBBB-2736-1G
4-(Heptyloxy)phenylboronsäure
4-(Heptyloxy)phenylboronsäure
Formel: C₁₃H₂₁BO₃
Molecular Weight: 236.12 g/mol
MDL: MFCD04039169
CAS-Nummer: 136370-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR