3-Chlor-4-(cyclopentylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Chloro-4-cyclopentylcarbamoylphenylboronic acid, (3-Chloro-4-(cyclopentylcarbamoyl)phenyl)boronic acid

BB-2739-5G BB-2739-1G
COBBBB-2739-5GEA 0 EUR
COBBBB-2739-5G COBBBB-2739-1G
3-Chlor-4-(cyclopentylcarbamoyl)phenylboronsäure
3-Chlor-4-(cyclopentylcarbamoyl)phenylboronsäure
Formel: C₁₂H₁₅BClNO₃
MDL: MFCD09258664
CAS-Nummer: 957061-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR