3,5-Dimethyl-4-propoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3,5-Dimethyl-4-propoxyphenyl)boronic acid

BB-2743-5G BB-2743-25G BB-2743-100G
COBBBB-2743-5GEA 98.3 EUR
COBBBB-2743-5G COBBBB-2743-25G COBBBB-2743-100G
3,5-Dimethyl-4-propoxyphenylboronsäure
3,5-Dimethyl-4-propoxyphenylboronsäure
Formel: C₁₁H₁₇BO₃
Molecular Weight: 208.0619 g/mol
MDL: MFCD05865178
CAS-Nummer: 357611-51-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR