3-Cyan-5-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

BB-2829-5G BB-2829-1G
COBBBB-2829-5GEA 0 EUR
COBBBB-2829-5G COBBBB-2829-1G
3-Cyan-5-nitrophenylboronsäure
3-Cyan-5-nitrophenylboronsäure
Formel: C₇H₅BN₂O₄
MDL: MFCD03411554
CAS-Nummer: 913835-33-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR