3-(Ethoxycarbonyl)-4-fluorphenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
3-Ethoxycarbonyl-4-fluorophenylboronic acid, (3-Ethoxycarbonyl-4-fluorophenyl)boronic acid
BB-2861-5G
BB-2861-1G
COBBBB-2861-5GEA
154
EUR
COBBBB-2861-5G
COBBBB-2861-1G
3-(Ethoxycarbonyl)-4-fluorphenylboronsäure
3-(Ethoxycarbonyl)-4-fluorphenylboronsäure
Formel:
C₉H₁₀BFO₄ Molecular Weight: 211.9827 g/mol |
MDL:
MFCD08061836 CAS-Nummer: 874219-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...