3-(Ethoxycarbonyl)-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Ethoxycarbonyl-4-fluorophenylboronic acid, (3-Ethoxycarbonyl-4-fluorophenyl)boronic acid

BB-2861-5G BB-2861-1G
COBBBB-2861-5GEA 154 EUR
COBBBB-2861-5G COBBBB-2861-1G
3-(Ethoxycarbonyl)-4-fluorphenylboronsäure
3-(Ethoxycarbonyl)-4-fluorphenylboronsäure
Formel: C₉H₁₀BFO₄
Molecular Weight: 211.9827 g/mol
MDL: MFCD08061836
CAS-Nummer: 874219-36-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR