4-Fluor-3-(methylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-Fluoro-3-(methylcarbamoyl)phenyl)boronic acid, 4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid

BB-2862-5G BB-2862-1G
COBBBB-2862-5GEA 1170 EUR
COBBBB-2862-5G COBBBB-2862-1G
4-Fluor-3-(methylcarbamoyl)phenylboronsäure
4-Fluor-3-(methylcarbamoyl)phenylboronsäure
Formel: C₈H₉BFNO₃
Molecular Weight: 196.97 g/mol
MDL: MFCD08235038
CAS-Nummer: 874219-19-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR