3-(Dimethylcarbamoyl)-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (3-(Dimethylcarbamoyl)-4-fluorophenyl)boronic acid, 3-Dimethylcarbamoyl-4-fluorobenzeneboronic acid, 4-Fluoro-3-(dimethylcarbamoyl)phenylboronic acid

BB-2870-5G BB-2870-1G
COBBBB-2870-5GEA 1580 EUR
COBBBB-2870-5G COBBBB-2870-1G
3-(Dimethylcarbamoyl)-4-fluorphenylboronsäure
3-(Dimethylcarbamoyl)-4-fluorphenylboronsäure
Formel: C₉H₁₁BFNO₃
Molecular Weight: 211.00 g/mol
MDL: MFCD08235046
CAS-Nummer: 874219-27-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR