3-Carboxy-4-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 3-Carboxy-4-fluorophenylboronic Acid, 3-Carboxy-4-fluorobenzeneboronic acid

BB-2879-25G BB-2879-1G BB-2879-5G
COBBBB-2879-25GEA 0 EUR
COBBBB-2879-25G COBBBB-2879-1G COBBBB-2879-5G
3-Carboxy-4-fluorphenylboronsäure
3-Carboxy-4-fluorphenylboronsäure
Formel: C₇H₆BFO₄
MDL: MFCD03095129
CAS-Nummer: 872460-12-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR