2-Fluor-(4-methoxycarbonyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 2-Fluoro-4-methoxycarbonylphenylboronic acid

BB-2888-5G BB-2888-25G BB-2888-1G
COBBBB-2888-5GEA 262 EUR
COBBBB-2888-5G COBBBB-2888-25G COBBBB-2888-1G
2-Fluor-(4-methoxycarbonyl)phenylboronsäure
2-Fluor-(4-methoxycarbonyl)phenylboronsäure
Formel: C₈H₈BFO₄
Molecular Weight: 197.96 g/mol
MDL: MFCD08436034
CAS-Nummer: 603122-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR